Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOS3
Entrez Gene ID
4846
Gene Name
nitric oxide synthase 3
Gene Aliases
ECNOS, eNOS
Chromosome Location
Chr.7: 150991056 - 151014599 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000603.4 NP_000594.2 4 - 5 712 73
NM_001160109.1 NP_001153581.1 3 - 4 416 73
NM_001160110.1 NP_001153582.1 3 - 4 416 73
NM_001160111.1 NP_001153583.1 3 - 4 416 73
XM_006716002.3 XP_006716065.1 3 - 4 469 73
XM_017012232.1 XP_016867721.1 3 - 4 469 73
XM_017012233.1 XP_016867722.1 3 - 4 368 73
XM_017012234.1 XP_016867723.1 3 - 4 469 73
GenBank mRNA AF400594.1 - 3 - 4 416 73
AK223636.1 - 3 - 4 436 73
AK225827.1 - 3 - 4 436 73
AK292928.1 - 3 - 4 469 73
AK302686.1 - 3 - 4 370 73
AK315213.1 - 3 - 4 438 73
BC063294.1 - 4 - 5 708 73
DQ256129.1 - 3 - 4 416 73
DQ256130.1 - 3 - 4 416 73
DQ256131.1 - 3 - 4 416 73
M93718.1 - 3 - 4 462 73
M95296.1 - 3 - 4 436 73

View Full Product Details