The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001795.1 | NP_001001795.1 | 1 - 2 | 317 | 91 |
GenBank mRNA | BC064373.1 | - | 1 - 2 | 317 | 91 |
BC064829.1 | - | 1 - 2 | 249 | 91 |