Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLN2
Entrez Gene ID
83660
Gene Name
talin 2
Gene Aliases
ILWEQ
Chromosome Location
Chr.15: 62390527 - 62844631 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015059.2 NP_055874.2 47 - 48 6475 98
XM_005254708.4 XP_005254765.1 50 - 51 6843 98
XM_005254710.4 XP_005254767.1 51 - 52 6981 98
XM_005254711.4 XP_005254768.1 50 - 51 7203 98
XM_005254712.4 XP_005254769.1 50 - 51 6860 98
XM_005254713.4 XP_005254770.1 50 - 51 7757 98
XM_005254714.3 XP_005254771.1 49 - 50 6746 98
XM_005254715.2 XP_005254772.1 49 - 50 7452 98
XM_006720717.3 XP_006720780.1 50 - 51 7577 98
XM_017022665.1 XP_016878154.1 50 - 51 7928 98
XM_017022666.1 XP_016878155.1 50 - 51 6787 98
XM_017022667.1 XP_016878156.1 48 - 49 6649 98
XM_017022668.1 XP_016878157.1 50 - 51 7576 98
XM_017022669.1 XP_016878158.1 49 - 50 7442 98
GenBank mRNA AB002318.2 - 33 - 34 4649 98
AB621808.1 - 28 - 29 3758 98
AF402000.1 - 47 - 48 6475 98
BX647654.1 - 18 - 19 4389 98

View Full Product Details