The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015059.2 | NP_055874.2 | 50 - 51 | 6882 | 158 |
XM_005254708.4 | XP_005254765.1 | 53 - 54 | 7250 | 158 | |
XM_005254710.4 | XP_005254767.1 | 54 - 55 | 7388 | 158 | |
XM_005254711.4 | XP_005254768.1 | 53 - 54 | 7610 | 158 | |
XM_005254712.4 | XP_005254769.1 | 53 - 54 | 7267 | 158 | |
XM_005254713.4 | XP_005254770.1 | 53 - 54 | 8164 | 158 | |
XM_005254714.3 | XP_005254771.1 | 52 - 53 | 7153 | 158 | |
XM_005254715.2 | XP_005254772.1 | 52 - 53 | 7859 | 158 | |
XM_006720717.3 | XP_006720780.1 | 53 - 54 | 7984 | 158 | |
XM_017022665.1 | XP_016878154.1 | 53 - 54 | 8335 | 158 | |
XM_017022666.1 | XP_016878155.1 | 53 - 54 | 7194 | 158 | |
XM_017022667.1 | XP_016878156.1 | 51 - 52 | 7056 | 158 | |
XM_017022668.1 | XP_016878157.1 | 53 - 54 | 7983 | 158 | |
XM_017022669.1 | XP_016878158.1 | 52 - 53 | 7849 | 158 | |
GenBank mRNA | AB002318.2 | - | 36 - 37 | 5056 | 158 |
AB621808.1 | - | 31 - 32 | 4165 | 158 | |
AF402000.1 | - | 50 - 51 | 6882 | 158 | |
BX647654.1 | - | 21 - 22 | 4796 | 158 |