Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location
Chr.22: 50343317 - 50445090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242898.1 NP_001229827.1 11 - 12 1723 125
NM_001242899.1 NP_001229828.1 11 - 12 1726 125
NM_001242900.1 NP_001229829.1 11 - 12 1723 125
NM_014678.4 NP_055493.2 11 - 12 1723 125
XM_005261960.3 XP_005262017.1 5 - 6 677 125
XM_006724431.2 XP_006724494.1 12 - 13 1834 125
XM_006724434.1 XP_006724497.1 13 - 14 1983 125
XM_006724435.2 XP_006724498.1 11 - 12 1708 125
XM_011530720.2 XP_011529022.1 10 - 11 1411 125
XM_011530721.2 XP_011529023.1 10 - 11 1408 125
XM_011530722.2 XP_011529024.1 10 - 11 1408 125
XM_011530723.2 XP_011529025.1 10 - 11 1411 125
XM_011530724.2 XP_011529026.1 10 - 11 1411 125
XM_011530726.2 XP_011529028.1 10 - 11 1411 125
XM_011530727.2 XP_011529029.1 10 - 11 1411 125
XM_011530728.2 XP_011529030.1 10 - 11 1411 125
XM_011530729.2 XP_011529031.1 10 - 11 1411 125
XM_011530730.2 XP_011529032.1 10 - 11 1408 125
XM_011530731.1 XP_011529033.1 10 - 11 1408 125
XM_011530732.2 XP_011529034.1 13 - 14 1887 125
XM_011530734.2 XP_011529036.1 12 - 13 1761 125
XM_011530735.2 XP_011529037.1 11 - 12 1711 125
XM_011530736.2 XP_011529038.1 12 - 13 1837 125
XM_011530737.2 XP_011529039.1 13 - 14 1986 125
XM_011530738.2 XP_011529040.1 12 - 13 1789 125
XM_011530739.2 XP_011529041.1 10 - 11 1451 125
XM_011530740.2 XP_011529042.1 12 - 13 1859 125
XM_017029116.1 XP_016884605.1 10 - 11 1408 125
XM_017029117.1 XP_016884606.1 14 - 15 2036 125
XM_017029118.1 XP_016884607.1 14 - 15 1988 125
XM_017029119.1 XP_016884608.1 13 - 14 1910 125
XM_017029120.1 XP_016884609.1 13 - 14 1884 125
XM_017029121.1 XP_016884610.1 11 - 12 1708 125
XM_017029122.1 XP_016884611.1 12 - 13 1786 125
XM_017029123.1 XP_016884612.1 11 - 12 1708 125
XM_017029124.1 XP_016884613.1 12 - 13 1834 125
XM_017029125.1 XP_016884614.1 11 - 12 1708 125
XM_017029126.1 XP_016884615.1 11 - 12 1711 125
XM_017029127.1 XP_016884616.1 12 - 13 1786 125
XM_017029128.1 XP_016884617.1 12 - 13 1834 125
XM_017029129.1 XP_016884618.1 11 - 12 1708 125
XM_017029130.1 XP_016884619.1 11 - 12 1708 125
XM_017029131.1 XP_016884620.1 12 - 13 1834 125
XM_017029132.1 XP_016884621.1 12 - 13 1786 125
XM_017029133.1 XP_016884622.1 11 - 12 1665 125
XM_017029134.1 XP_016884623.1 11 - 12 1662 125
XM_017029135.1 XP_016884624.1 12 - 13 1712 125
XM_017029136.1 XP_016884625.1 11 - 12 1662 125
XM_017029137.1 XP_016884626.1 11 - 12 1662 125
XM_017029138.1 XP_016884627.1 11 - 12 1662 125
GenBank mRNA AB014585.1 - 11 - 12 1695 125
AK002063.1 - 4 - 5 519 125
AK022669.1 - 1 - 2 227 125
AK302472.1 - 11 - 12 1634 125
BC000976.4 - 12 - 13 1712 125
BC006568.2 - 11 - 12 1709 125
BC032664.1 - 11 - 12 1723 125
BC041698.1 - 11 - 12 1693 125
BC052995.2 - 11 - 12 1629 125

View Full Product Details