The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177984.2 | NP_001171455.1 | 25 - 26 | 4854 | 152 |
NM_014191.3 | NP_055006.1 | 26 - 27 | 4977 | 152 | |
XM_006719556.3 | XP_006719619.1 | 26 - 27 | 5568 | 152 | |
XM_011538650.2 | XP_011536952.1 | 26 - 27 | 4996 | 152 | |
XM_011538651.2 | XP_011536953.1 | 26 - 27 | 4961 | 152 | |
XM_017019794.1 | XP_016875283.1 | 26 - 27 | 4974 | 152 | |
XM_017019795.1 | XP_016875284.1 | 25 - 26 | 4870 | 152 | |
GenBank mRNA | AB027567.1 | - | 26 - 27 | 4977 | 152 |
AF225988.1 | - | 25 - 26 | 4799 | 152 | |
FJ611941.1 | - | 24 - 25 | 4676 | 152 |