The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048210.2 | NP_001041675.1 | 5 - 6 | 702 | 136 |
NM_015127.4 | NP_055942.1 | 5 - 6 | 562 | 136 | |
GenBank mRNA | AB018304.2 | - | 4 - 5 | 563 | 136 |
AB052915.1 | - | 4 - 5 | 573 | 136 | |
AB052916.1 | - | 4 - 5 | 423 | 136 | |
AK090563.1 | - | 6 - 7 | 824 | 136 | |
AL550290.3 | - | 5 - 6 | 670 | 136 | |
BC002939.2 | - | 5 - 6 | 520 | 136 |