Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXLNA
Entrez Gene ID
200081
Gene Name
taxilin alpha
Gene Aliases
IL14, TXLN
Chromosome Location
Chr.1: 32179669 - 32198285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175852.3 NP_787048.1 4 - 5 660 115
XM_017000561.1 XP_016856050.1 3 - 4 1052 115
XM_017000562.1 XP_016856051.1 3 - 4 953 115
XM_017000563.1 XP_016856052.1 4 - 5 834 115
XM_017000564.1 XP_016856053.1 3 - 4 514 115
XM_017000565.1 XP_016856054.1 3 - 4 608 115
GenBank mRNA AF516206.1 - 3 - 4 673 115
AL832338.1 - 4 - 5 697 115
AL832636.2 - 4 - 5 719 115
AL832637.2 - 3 - 4 876 115
BC080578.1 - 3 - 4 637 115
BC103823.1 - 3 - 4 625 115
BC103824.1 - 3 - 4 625 115

View Full Product Details