Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
Chromosome Location
Chr.19: 44503838 - 44530100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102597.2 NP_001096067.2 5 - 6 1049 134
NM_001102600.2 NP_001096070.2 5 - 6 1049 134
XM_011526429.2 XP_011524731.1 6 - 7 1703 134
XM_011526430.2 XP_011524732.1 6 - 7 1704 134
XM_011526431.2 XP_011524733.1 6 - 7 1705 134
XM_011526432.2 XP_011524734.1 2 - 3 367 134
GenBank mRNA AY358129.1 - 5 - 6 1312 134
EU008600.1 - 5 - 6 1049 134
EU008603.1 - 5 - 6 1049 134

View Full Product Details