The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102597.2 | NP_001096067.2 | 6 - 7 | 1326 | 147 |
NM_001102600.2 | NP_001096070.2 | 6 - 7 | 1326 | 147 | |
XM_011526429.2 | XP_011524731.1 | 7 - 8 | 1980 | 147 | |
XM_011526430.2 | XP_011524732.1 | 7 - 8 | 1981 | 147 | |
XM_011526431.2 | XP_011524733.1 | 7 - 8 | 1982 | 147 | |
XM_011526432.2 | XP_011524734.1 | 3 - 4 | 644 | 147 | |
GenBank mRNA | AY358129.1 | - | 6 - 7 | 1589 | 147 |
EU008600.1 | - | 6 - 7 | 1326 | 147 | |
EU008603.1 | - | 6 - 7 | 1326 | 147 |