The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002806.3 | NP_002797.3 | 1 - 2 | 127 | 180 |
GenBank mRNA | BM788029.1 | - | 1 - 2 | 127 | 180 |