The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173808.2 | NP_776169.2 | 2 - 3 | 507 | 112 |
XM_011541200.2 | XP_011539502.1 | 2 - 3 | 763 | 112 | |
XM_011541201.2 | XP_011539503.1 | 2 - 3 | 763 | 112 | |
XM_017000961.1 | XP_016856450.1 | 2 - 3 | 763 | 112 | |
GenBank mRNA | AK092307.1 | - | 2 - 3 | 414 | 112 |
AK295481.1 | - | 2 - 3 | 405 | 112 | |
BC036771.1 | - | 2 - 3 | 429 | 112 | |
BX538014.1 | - | 2 - 3 | 411 | 112 |