The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271807.1 | NP_001258736.1 | 1 - 2 | 100 | 173 |
NM_178174.3 | NP_835468.1 | 1 - 2 | 100 | 173 | |
XM_017010822.1 | XP_016866311.1 | 3 - 4 | 290 | 173 | |
XM_017010823.1 | XP_016866312.1 | 2 - 3 | 178 | 173 | |
XM_017010824.1 | XP_016866313.1 | 2 - 3 | 184 | 173 | |
XM_017010825.1 | XP_016866314.1 | 3 - 4 | 290 | 173 | |
GenBank mRNA | AY358357.1 | - | 1 - 2 | 88 | 173 |