The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152344.3 | NP_689557.1 | 2 - 3 | 442 | 79 |
XM_011524309.2 | XP_011522611.1 | 1 - 2 | 206 | 79 | |
GenBank mRNA | AK055218.1 | - | 1 - 2 | 170 | 79 |
AK300637.1 | - | 1 - 2 | 128 | 79 | |
AL832076.1 | - | 1 - 1 | 131 | 79 | |
BC044587.1 | - | 1 - 2 | 170 | 79 |