The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030629.2 | NP_085132.1 | 17 - 18 | 1788 | 139 |
NM_198390.2 | NP_938204.2 | 17 - 18 | 2014 | 139 | |
XM_005256179.4 | XP_005256236.4 | 17 - 18 | 2969 | 139 | |
XM_005256181.2 | XP_005256238.1 | 16 - 17 | 1745 | 139 | |
XM_005256182.1 | XP_005256239.1 | 15 - 16 | 1502 | 139 | |
XM_011523353.1 | XP_011521655.1 | 15 - 16 | 1882 | 139 | |
XM_017023733.1 | XP_016879222.1 | 15 - 16 | 6359 | 139 | |
GenBank mRNA | AB051481.1 | - | 17 - 18 | 1895 | 139 |
AK096598.1 | - | 17 - 18 | 1975 | 139 | |
AK304418.1 | - | 16 - 17 | 1750 | 139 | |
AY172689.1 | - | 17 - 18 | 1788 | 139 | |
BC038113.1 | - | 15 - 16 | 1711 | 139 |