The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015111.1 | NP_055926.1 | 2 - 3 | 584 | 143 |
XM_006714834.3 | XP_006714897.1 | 2 - 3 | 614 | 143 | |
XM_011534473.1 | XP_011532775.1 | 2 - 3 | 633 | 143 | |
XM_011534474.1 | XP_011532776.1 | 2 - 3 | 633 | 143 | |
GenBank mRNA | AB002339.1 | - | 1 - 2 | 345 | 143 |
BC053323.1 | - | 2 - 3 | 584 | 143 |