The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022170.1 | NP_071496.1 | 2 - 3 | 257 | 96 |
NM_031992.1 | NP_114381.1 | 2 - 3 | 257 | 96 | |
XM_011516556.1 | XP_011514858.1 | 2 - 3 | 290 | 96 | |
XM_011516557.1 | XP_011514859.1 | 2 - 3 | 275 | 96 | |
GenBank mRNA | AK290676.1 | - | 2 - 3 | 277 | 96 |
AK297651.1 | - | 2 - 3 | 302 | 96 | |
AK314749.1 | - | 2 - 3 | 359 | 96 | |
BC010021.1 | - | 1 - 1 | 255 | 96 | |
BC021214.2 | - | 2 - 3 | 255 | 96 | |
BC066928.1 | - | 1 - 1 | 274 | 96 | |
D26068.1 | - | 2 - 3 | 257 | 96 |