Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR92
Entrez Gene ID
116143
Gene Name
WD repeat domain 92
Gene Aliases
-
Chromosome Location
Chr.2: 68130149 - 68157560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256476.1 NP_001243405.1 5 - 6 746 154
NM_138458.3 NP_612467.1 5 - 6 746 154
NR_046234.1 5 - 6 619 154
GenBank mRNA AK056303.1 - 5 - 6 710 154
AK058090.1 - 5 - 6 619 154
AL834142.1 - 5 - 6 582 154
AY541587.1 - 5 - 6 721 154
BC014022.2 - 5 - 6 668 154
BC020271.1 - 5 - 6 700 154
BC036904.1 - 5 - 6 556 154
BC066657.1 - 5 - 6 696 154

View Full Product Details