The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270546.1 | NP_001257475.1 | 19 - 20 | 2832 | 125 |
NM_006738.5 | NP_006729.4 | 27 - 28 | 7033 | 125 | |
NM_007200.4 | NP_009131.2 | 27 - 28 | 7021 | 125 | |
GenBank mRNA | AB055890.1 | - | 28 - 29 | 7040 | 125 |
AB209414.1 | - | 16 - 17 | 2314 | 125 | |
AF126008.1 | - | 19 - 20 | 2675 | 125 | |
AF387101.1 | - | 26 - 27 | 6827 | 125 | |
AF406992.1 | - | 27 - 28 | 6920 | 125 | |
AK309805.1 | - | 19 - 20 | 2701 | 125 | |
BC063592.1 | - | 14 - 15 | 1808 | 125 | |
BC140862.1 | - | 12 - 13 | 1644 | 125 | |
BC171798.1 | - | 12 - 13 | 1644 | 125 |