The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270546.1 | NP_001257475.1 | 22 - 23 | 3288 | 73 |
NM_006738.5 | NP_006729.4 | 30 - 31 | 7489 | 73 | |
NM_007200.4 | NP_009131.2 | 30 - 31 | 7477 | 73 | |
GenBank mRNA | AB055890.1 | - | 31 - 32 | 7496 | 73 |
AB209414.1 | - | 19 - 20 | 2770 | 73 | |
AF126008.1 | - | 22 - 23 | 3131 | 73 | |
AF387101.1 | - | 29 - 30 | 7283 | 73 | |
AF406992.1 | - | 30 - 31 | 7376 | 73 | |
BC063592.1 | - | 17 - 18 | 2264 | 73 | |
BC140862.1 | - | 15 - 16 | 2100 | 73 | |
BC171798.1 | - | 15 - 16 | 2100 | 73 |