The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025234.1 | NP_001020405.1 | 4 - 5 | 479 | 159 |
NM_001025235.1 | NP_001020406.1 | 4 - 5 | 478 | 159 | |
NM_001025236.1 | NP_001020407.1 | 4 - 5 | 483 | 159 | |
NM_001025237.1 | NP_001020408.1 | 4 - 5 | 515 | 159 | |
NM_001025238.1 | NP_001020409.1 | 3 - 4 | 365 | 159 | |
NM_001025239.1 | NP_001020410.1 | 3 - 4 | 385 | 159 | |
NM_003271.4 | NP_003262.1 | 4 - 5 | 465 | 159 | |
XM_005253102.1 | XP_005253159.1 | 3 - 4 | 384 | 159 | |
XM_011520336.1 | XP_011518638.1 | 3 - 4 | 824 | 159 | |
XM_011520337.2 | XP_011518639.1 | 3 - 4 | 686 | 159 | |
XM_011520338.1 | XP_011518640.1 | 3 - 4 | 406 | 159 | |
XM_011520340.2 | XP_011518642.1 | 3 - 4 | 464 | 159 | |
XM_011520341.2 | XP_011518643.1 | 3 - 4 | 385 | 159 | |
GenBank mRNA | AF022813.1 | - | 3 - 4 | 360 | 159 |
AF054841.1 | - | 3 - 4 | 374 | 159 | |
AK291089.1 | - | 4 - 5 | 440 | 159 | |
BC000389.2 | - | 4 - 5 | 391 | 159 | |
BC019314.2 | - | 3 - 4 | 376 | 159 | |
BC093031.1 | - | 4 - 5 | 434 | 159 | |
BG752017.1 | - | 4 - 5 | 472 | 159 | |
BI820547.1 | - | 4 - 5 | 535 | 159 | |
BQ679002.1 | - | - | 160 | ||
BT006776.1 | - | 2 - 3 | 256 | 159 | |
CR457108.1 | - | 2 - 3 | 256 | 159 |