The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308297.1 | NP_001295226.1 | 14 - 15 | 2094 | 111 |
NM_012411.5 | NP_036543.4 | 13 - 14 | 2001 | 111 | |
NM_015967.6 | NP_057051.3 | 15 - 16 | 2166 | 111 | |
XM_011541221.1 | XP_011539523.1 | 14 - 15 | 2069 | 111 | |
XM_011541222.1 | XP_011539524.1 | 15 - 16 | 2147 | 111 | |
XM_011541225.2 | XP_011539527.1 | 14 - 15 | 2096 | 111 | |
XM_017001004.1 | XP_016856493.1 | 15 - 16 | 2147 | 111 | |
XM_017001005.1 | XP_016856494.1 | 15 - 16 | 2128 | 111 | |
GenBank mRNA | AF001846.1 | - | 15 - 16 | 2074 | 111 |
AF077031.1 | - | 15 - 16 | 2102 | 111 | |
AF150732.1 | - | 15 - 16 | 2076 | 111 | |
AK303124.1 | - | 11 - 12 | 1801 | 111 | |
AK310570.1 | - | 10 - 11 | 1738 | 111 | |
AK310698.1 | - | 15 - 16 | 2119 | 111 | |
BC071670.1 | - | 13 - 14 | 1940 | 111 | |
JN084012.1 | - | 14 - 15 | 1971 | 111 |