Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATXN7L2
Entrez Gene ID
127002
Gene Name
ataxin 7 like 2
Gene Aliases
-
Chromosome Location
Chr.1: 109483904 - 109492804 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270445.3 XP_005270502.1 11 - 12 2380 101
XM_011540638.2 XP_011538940.1 10 - 11 2279 101
XM_011540639.2 XP_011538941.1 11 - 12 3649 101
XM_011540641.2 XP_011538943.1 11 - 12 2982 101
XM_011540644.2 XP_011538946.1 13 - 14 2464 101
XM_011540645.1 XP_011538947.1 10 - 11 2225 101
XM_017000267.1 XP_016855756.1 12 - 13 3111 101
XM_017000268.1 XP_016855757.1 12 - 13 2444 101
GenBank mRNA AK090460.1 - 7 - 8 4236 101

View Full Product Details