The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003214.3 | NP_003205.2 | 10 - 11 | 1136 | 125 |
GenBank mRNA | AF142482.1 | - | 10 - 11 | 1090 | 125 |
AK226144.1 | - | 6 - 7 | 2496 | 125 | |
BC010053.2 | - | 10 - 11 | 992 | 125 | |
BC027877.1 | - | 8 - 9 | 874 | 125 | |
BC091488.1 | - | 10 - 11 | 1061 | 125 | |
X94439.1 | - | 9 - 10 | 900 | 125 |