The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015692.2 | NP_056507.2 | 38 - 39 | 5128 | 170 |
XM_011527917.1 | XP_011526219.1 | 38 - 39 | 5095 | 170 | |
XM_011527918.1 | XP_011526220.1 | 38 - 39 | 5131 | 170 | |
XM_011527919.1 | XP_011526221.1 | 38 - 39 | 5131 | 170 | |
XM_011527920.2 | XP_011526222.1 | 38 - 39 | 5131 | 170 | |
XM_011527921.2 | XP_011526223.1 | 38 - 39 | 5131 | 170 | |
XM_011527922.1 | XP_011526224.1 | 38 - 39 | 5131 | 170 | |
XM_011527923.1 | XP_011526225.1 | 38 - 39 | 5131 | 170 | |
XM_011527924.2 | XP_011526226.1 | 38 - 39 | 5131 | 170 | |
XM_011527925.1 | XP_011526227.1 | 22 - 23 | 2973 | 170 | |
XM_017026594.1 | XP_016882083.1 | 38 - 39 | 5131 | 170 |