The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018947.5 | NP_061820.1 | 2 - 3 | 343 | 85 |
GenBank mRNA | AK311836.1 | - | 2 - 3 | 274 | 85 |
AL713681.1 | - | 2 - 3 | 233 | 85 | |
BC005299.1 | - | 2 - 3 | 240 | 85 | |
BC008475.1 | - | 2 - 3 | 242 | 85 | |
BC008477.1 | - | 2 - 3 | 242 | 85 | |
BC009578.1 | - | 2 - 3 | 242 | 85 | |
BC009579.1 | - | 2 - 3 | 242 | 85 | |
BC009582.1 | - | 2 - 3 | 242 | 85 | |
BC009587.1 | - | 2 - 3 | 242 | 85 | |
BC009602.1 | - | 2 - 3 | 242 | 85 | |
BC009607.1 | - | 2 - 3 | 246 | 85 | |
BC014359.1 | - | 2 - 3 | 246 | 85 | |
BC014361.1 | - | 2 - 3 | 246 | 85 | |
BC015130.1 | - | 2 - 3 | 246 | 85 | |
BC016006.1 | - | 2 - 3 | 246 | 85 | |
BC021994.1 | - | 2 - 3 | 246 | 85 | |
BC022330.1 | - | 2 - 3 | 242 | 85 | |
BC024216.1 | - | 2 - 3 | 285 | 85 | |
BC067222.1 | - | 2 - 3 | 85 | ||
BC068464.1 | - | 2 - 3 | 242 | 85 | |
BC070156.1 | - | 2 - 3 | 242 | 85 | |
BC070346.1 | - | 2 - 3 | 243 | 85 | |
BC071761.1 | - | 2 - 3 | 242 | 85 | |
BT006946.1 | - | 1 - 2 | 173 | 85 | |
BU597562.1 | - | 2 - 3 | 321 | 85 | |
CR542175.1 | - | 1 - 2 | 173 | 85 | |
DB447825.1 | - | 2 - 3 | 343 | 85 |