Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD53
Entrez Gene ID
79998
Gene Name
ankyrin repeat domain 53
Gene Aliases
-
Chromosome Location
Chr.2: 70977183 - 70985499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115116.1 NP_001108588.1 4 - 5 984 131
NM_024933.3 NP_079209.3 4 - 5 984 131
XM_005264569.4 XP_005264626.1 4 - 5 2144 131
GenBank mRNA AK093479.1 - 4 - 5 820 131
BC035234.1 - 4 - 5 984 131

View Full Product Details