Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
Chromosome Location
Chr.12: 112418732 - 112509913 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002834.3 NP_002825.3 4 - 5 907 133
NM_080601.1 NP_542168.1 4 - 5 907 133
XM_006719526.2 XP_006719589.1 4 - 5 685 133
XM_011538613.2 XP_011536915.1 4 - 5 721 133
XM_017019722.1 XP_016875211.1 4 - 5 724 133
GenBank mRNA AK289854.1 - 4 - 5 685 133
BC008692.1 - 4 - 5 730 133
BT007106.1 - 3 - 4 527 133
D13540.1 - 4 - 5 683 133
EU779706.1 - 1 - 2 194 133
EU779708.1 - 3 - 4 521 133
L03535.1 - 4 - 5 640 133
L07527.1 - 4 - 5 680 133
L08807.1 - 4 - 5 725 133
X70766.1 - 4 - 5 656 133

View Full Product Details