Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP40
Entrez Gene ID
343578
Gene Name
Rho GTPase activating protein 40
Gene Aliases
C20orf95, dJ1100H13.4
Chromosome Location
Chr.20: 38601934 - 38650652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164431.1 NP_001157903.1 8 - 9 1128 104

View Full Product Details