The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007538.1 | NP_001007539.1 | 1 - 2 | 420 | 106 |
GenBank mRNA | AY358775.1 | - | 1 - 2 | 408 | 106 |
BC132898.1 | - | 1 - 2 | 387 | 106 | |
BC137092.1 | - | 1 - 2 | 387 | 106 |