Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RABL6
- Entrez Gene ID
-
55684
- Gene Name
- RAB, member RAS oncogene family-like 6
- Gene Aliases
-
C9orf86, PARF, RBEL1, pp8875
- Chromosome Location
-
Chr.9: 136807922 - 136841187 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK094382.1 |
- |
1 - 2 |
726 |
145 |
View Full Product Details