Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZGP1P1
Entrez Gene ID
646282
Gene Name
alpha-2-glycoprotein 1, zinc-binding pseudogene 1
Gene Aliases
-
Chromosome Location
Chr.7: 99980762 - 99984237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036679.1 1 - 2 87 120
GenBank mRNA AK307369.1 - 1 - 2 87 120

View Full Product Details