The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199085.1 | NP_001186014.1 | 15 - 16 | 3017 | 144 |
NM_001199089.1 | NP_001186018.1 | 15 - 16 | 2730 | 144 | |
NM_001199091.1 | NP_001186020.1 | 14 - 15 | 2696 | 144 | |
NM_001199092.1 | NP_001186021.1 | 11 - 12 | 1540 | 144 | |
NM_173533.3 | NP_775804.2 | 14 - 15 | 2855 | 144 | |
XM_005244934.1 | XP_005244991.1 | 15 - 16 | 2802 | 144 | |
XM_005244935.4 | XP_005244992.1 | 15 - 16 | 2707 | 144 | |
XM_017000473.1 | XP_016855962.1 | 14 - 15 | 2561 | 144 | |
GenBank mRNA | AK092142.1 | - | 11 - 12 | 1553 | 144 |
AK126889.1 | - | 14 - 15 | 2696 | 144 | |
BC043368.1 | - | 6 - 7 | 867 | 144 | |
BC089387.1 | - | 12 - 13 | 1757 | 144 | |
BC130532.1 | - | 15 - 16 | 2694 | 144 | |
BC144058.1 | - | 15 - 16 | 2749 | 144 |