The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304829.1 | NP_001291758.1 | 12 - 13 | 1747 | 130 |
NM_194292.2 | NP_919268.1 | 13 - 14 | 1827 | 130 | |
XM_017000486.1 | XP_016855975.1 | 13 - 14 | 1593 | 130 | |
GenBank mRNA | AK025750.1 | - | 1 - 2 | 125 | 130 |
AK302513.1 | - | 14 - 15 | 1577 | 130 | |
AK307824.1 | - | 12 - 13 | 1613 | 130 | |
AL834265.1 | - | 9 - 10 | 1090 | 130 | |
AY359522.1 | - | 13 - 14 | 1687 | 130 | |
BC101026.3 | - | 12 - 13 | 1540 | 130 | |
BX641109.1 | - | 12 - 13 | 1738 | 130 | |
BX648406.1 | - | 13 - 14 | 1584 | 130 |