The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043318.2 | NP_001036783.1 | 6 - 7 | 2707 | 147 |
NM_194278.3 | NP_919254.2 | 6 - 7 | 3086 | 147 | |
XM_005268204.1 | XP_005268261.1 | 6 - 7 | 3086 | 147 | |
XM_005268205.1 | XP_005268262.1 | 6 - 7 | 2642 | 147 | |
XM_005268206.1 | XP_005268263.1 | 7 - 8 | 2859 | 147 | |
GenBank mRNA | AK090424.1 | - | 1 - 2 | 2344 | 147 |
AK127785.1 | - | 7 - 8 | 3085 | 147 | |
BC003121.1 | - | 6 - 7 | 1879 | 147 | |
BC006451.1 | - | 6 - 7 | 1541 | 147 | |
BC006511.2 | - | 6 - 7 | 2337 | 147 | |
BC009202.2 | - | 7 - 8 | 2707 | 147 | |
BC015668.2 | - | 6 - 7 | 2720 | 147 | |
BC025330.1 | - | 6 - 7 | 2337 | 147 |