Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIR99AHG
Entrez Gene ID
388815
Gene Name
mir-99a-let-7c cluster host gene
Gene Aliases
C21orf34, C21orf35, LINC00478, MONC
Chromosome Location
Chr.21: 16070488 - 16631727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027790.3 5 - 6 517 165
GenBank mRNA AK095614.1 - 5 - 6 517 165

View Full Product Details