The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244606.1 | NP_001231535.1 | 3 - 4 | 264 | 127 |
NM_032549.3 | NP_115938.1 | 2 - 3 | 578 | 127 | |
XM_005250630.3 | XP_005250687.1 | 4 - 5 | 346 | 127 | |
XM_011516605.2 | XP_011514907.1 | 3 - 4 | 278 | 127 | |
XM_011516608.2 | XP_011514910.1 | 2 - 3 | 540 | 127 | |
XM_011516609.2 | XP_011514911.1 | 2 - 3 | 540 | 127 | |
XM_011516611.2 | XP_011514913.1 | 2 - 3 | 540 | 127 | |
XM_011516613.2 | XP_011514915.1 | 2 - 3 | 540 | 127 | |
XM_017012699.1 | XP_016868188.1 | 2 - 3 | 540 | 127 | |
XM_017012700.1 | XP_016868189.1 | 3 - 4 | 278 | 127 | |
XM_017012701.1 | XP_016868190.1 | 3 - 4 | 229 | 127 | |
XM_017012702.1 | XP_016868191.1 | 4 - 5 | 346 | 127 | |
XM_017012703.1 | XP_016868192.1 | 2 - 3 | 540 | 127 | |
XM_017012704.1 | XP_016868193.1 | 2 - 3 | 540 | 127 | |
XM_017012705.1 | XP_016868194.1 | 3 - 4 | 316 | 127 | |
GenBank mRNA | AF359563.1 | - | 2 - 3 | 578 | 127 |
AK311968.1 | - | 1 - 2 | 229 | 127 | |
BC008497.1 | - | 3 - 4 | 229 | 127 |