The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006712117.3 | XP_006712180.1 | 4 - 5 | 364 | 146 |
XM_006712118.3 | XP_006712181.1 | 4 - 5 | 364 | 146 | |
XM_011533133.2 | XP_011531435.1 | 4 - 5 | 432 | 146 | |
XM_017005111.1 | XP_016860600.1 | 4 - 5 | 364 | 146 |