The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194313.2 | NP_919289.2 | 11 - 12 | 3993 | 137 |
XM_005251451.2 | XP_005251508.1 | 7 - 8 | 2886 | 137 | |
XM_006716764.2 | XP_006716827.1 | 5 - 6 | 2597 | 137 | |
XM_011517861.1 | XP_011516163.1 | 12 - 13 | 4214 | 137 | |
XM_011517863.2 | XP_011516165.1 | 11 - 12 | 5303 | 137 | |
XM_017014697.1 | XP_016870186.1 | 11 - 12 | 4307 | 137 | |
XM_017014698.1 | XP_016870187.1 | 10 - 11 | 4080 | 137 | |
GenBank mRNA | AK125180.1 | - | 1 - 2 | 707 | 137 |
AK125872.1 | - | 2 - 3 | 2369 | 137 | |
BC048311.1 | - | 2 - 3 | 740 | 137 | |
BC110502.1 | - | 2 - 3 | 2351 | 137 | |
BC110503.1 | - | 2 - 3 | 2351 | 137 |