The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077594.1 | NP_001071062.1 | 7 - 8 | 1661 | 123 |
XM_011537323.2 | XP_011535625.1 | 8 - 9 | 2336 | 123 | |
XM_011537324.2 | XP_011535626.1 | 8 - 9 | 2320 | 123 | |
XM_011537325.2 | XP_011535627.1 | 9 - 10 | 2092 | 123 | |
XM_011537327.2 | XP_011535629.1 | 8 - 9 | 1791 | 123 | |
XM_011537328.2 | XP_011535630.1 | 9 - 10 | 1823 | 123 | |
XM_011537329.2 | XP_011535631.1 | 9 - 10 | 2231 | 123 | |
XM_011537330.2 | XP_011535632.1 | 10 - 11 | 2012 | 123 | |
XM_011537331.1 | XP_011535633.1 | 9 - 10 | 1807 | 123 | |
XM_011537332.2 | XP_011535634.1 | 8 - 9 | 2036 | 123 | |
XM_011537333.2 | XP_011535635.1 | 9 - 10 | 2068 | 123 | |
XM_011537334.1 | XP_011535636.1 | 7 - 8 | 2019 | 123 | |
GenBank mRNA | AK000671.1 | - | 4 - 5 | 1433 | 123 |
BC113655.1 | - | 7 - 8 | 1661 | 123 | |
BC117375.1 | - | 7 - 8 | 1661 | 123 |