The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113482.1 | NP_001106954.1 | 3 - 4 | 820 | 114 |
GenBank mRNA | AK301399.1 | - | 3 - 4 | 414 | 114 |
BC063587.1 | - | 3 - 4 | 277 | 114 |