Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
Chromosome Location
Chr.1: 33513999 - 34165842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281956.1 NP_001268885.1 36 - 37 5896 115
NM_052896.4 NP_443128.2 36 - 37 5628 115
XM_017000185.1 XP_016855674.1 36 - 37 5890 115
XM_017000186.1 XP_016855675.1 36 - 37 5890 115
XM_017000187.1 XP_016855676.1 36 - 37 5719 115
XM_017000188.1 XP_016855677.1 36 - 37 5628 115
XM_017000189.1 XP_016855678.1 36 - 37 5890 115
XM_017000190.1 XP_016855679.1 30 - 31 5155 115
XM_017000191.1 XP_016855680.1 36 - 37 5890 115
XM_017000192.1 XP_016855681.1 36 - 37 5890 115
XM_017000193.1 XP_016855682.1 36 - 37 5890 115
XM_017000194.1 XP_016855683.1 36 - 37 5628 115
GenBank mRNA AB212622.1 - 36 - 37 5896 115
AK122603.1 - 4 - 5 522 115
AK127722.1 - 15 - 16 2560 115
AK291019.1 - 15 - 16 2401 115
AY210418.1 - 36 - 37 5628 115

View Full Product Details