Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD3
Entrez Gene ID
89846
Gene Name
FYVE, RhoGEF and PH domain containing 3
Gene Aliases
ZFYVE5
Chromosome Location
Chr.9: 92947319 - 93036236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083536.1 NP_001077005.1 14 - 15 2182 115
NM_001286993.1 NP_001273922.1 14 - 15 2164 115
NM_033086.2 NP_149077.2 14 - 15 2082 115
XM_017015274.1 XP_016870763.1 14 - 15 1924 115
GenBank mRNA AK000004.1 - 13 - 14 1759 115
AK225843.1 - 14 - 15 1834 115
AK292676.1 - 14 - 15 2164 115
AK301974.1 - 6 - 7 538 115
AY211386.1 - 14 - 15 2018 115
BC032429.1 - 14 - 15 1901 115
BC111054.1 - 14 - 15 2019 115
BX647829.1 - 14 - 15 2037 115

View Full Product Details