Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD3
Entrez Gene ID
89846
Gene Name
FYVE, RhoGEF and PH domain containing 3
Gene Aliases
ZFYVE5
Chromosome Location
Chr.9: 92947319 - 93036236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083536.1 NP_001077005.1 17 - 18 2552 110
NM_001286993.1 NP_001273922.1 17 - 18 2531 110
NM_033086.2 NP_149077.2 17 - 18 2452 110
XM_017015274.1 XP_016870763.1 17 - 18 2294 110
GenBank mRNA AK000004.1 - 16 - 17 3773 110
AK225843.1 - 17 - 18 2204 110
AK292676.1 - 17 - 18 2531 110
AK301974.1 - 9 - 10 908 110
AY211386.1 - 16 - 17 4055 110
BC032429.1 - 17 - 18 2271 110
BC111054.1 - 17 - 18 2389 110
BX647829.1 - 17 - 18 2407 110

View Full Product Details