The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201365.1 | NP_001188294.1 | 3 - 4 | 397 | 149 |
NM_004290.4 | NP_004281.1 | 3 - 4 | 611 | 149 | |
NM_183399.2 | NP_899646.1 | 2 - 3 | 223 | 149 | |
NM_183400.2 | NP_899647.1 | 2 - 3 | 437 | 149 | |
NM_183401.2 | NP_899648.1 | 3 - 4 | 417 | 149 | |
XM_005268536.2 | XP_005268593.1 | 3 - 4 | 714 | 149 | |
XM_005268537.2 | XP_005268594.1 | 3 - 4 | 758 | 149 | |
XM_005268542.2 | XP_005268599.1 | 3 - 4 | 454 | 149 | |
XM_005268543.4 | XP_005268600.1 | 2 - 3 | 260 | 149 | |
XM_011537714.2 | XP_011536016.1 | 2 - 3 | 228 | 149 | |
XM_017010070.1 | XP_016865559.1 | 3 - 4 | 362 | 149 | |
XM_017010073.1 | XP_016865562.1 | 3 - 4 | 492 | 149 | |
XM_017010074.1 | XP_016865563.1 | 3 - 4 | 758 | 149 | |
XM_017010075.1 | XP_016865564.1 | 3 - 4 | 417 | 149 | |
XM_017010076.1 | XP_016865565.1 | 3 - 4 | 403 | 149 | |
GenBank mRNA | AB022663.1 | - | 3 - 4 | 385 | 149 |
AF060544.1 | - | 2 - 3 | 188 | 149 | |
AK023698.1 | - | 2 - 3 | 187 | 149 | |
AK023884.1 | - | 3 - 4 | 353 | 149 | |
AK294262.1 | - | 3 - 4 | 356 | 149 | |
BC126185.1 | - | 3 - 4 | 356 | 149 | |
BC144061.1 | - | 3 - 4 | 356 | 149 | |
BX647955.1 | - | 3 - 4 | 384 | 149 | |
CB997918.1 | - | 2 - 3 | 245 | 149 | |
CD106708.1 | - | 3 - 4 | 423 | 149 | |
CR456702.1 | - | 1 - 2 | 149 | 149 | |
DC358984.1 | - | 3 - 4 | 417 | 149 | |
DC396586.1 | - | 3 - 4 | 397 | 149 |