The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286246.1 | NP_001273175.1 | 2 - 3 | 293 | 140 |
NM_178502.3 | NP_848597.1 | 3 - 4 | 454 | 140 | |
XM_005268703.1 | XP_005268760.1 | 3 - 4 | 629 | 140 | |
XM_011538022.2 | XP_011536324.1 | 2 - 3 | 832 | 140 | |
GenBank mRNA | AK092085.1 | - | 3 - 4 | 255 | 140 |
AK128752.1 | - | 1 - 2 | 1422 | 140 | |
BC114441.1 | - | 2 - 3 | 183 | 140 | |
BC114498.1 | - | 2 - 3 | 183 | 140 | |
DA684401.1 | - | 2 - 3 | 293 | 140 |