The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178439.4 | NP_848526.1 | 1 - 2 | 545 | 171 |
XM_011533033.1 | XP_011531335.1 | 1 - 2 | 509 | 171 | |
XM_011533034.2 | XP_011531336.1 | 1 - 2 | 491 | 171 | |
XM_017004704.1 | XP_016860193.1 | 1 - 2 | 444 | 171 | |
GenBank mRNA | AK023119.1 | - | 1 - 2 | 451 | 171 |
AK024042.1 | - | 1 - 2 | 509 | 171 | |
BC007420.2 | - | 1 - 2 | 465 | 171 | |
BI550798.1 | - | 1 - 2 | 550 | 171 |