The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153810.4 | NP_722517.3 | 4 - 5 | 1163 | 83 |
GenBank mRNA | AJ544771.1 | - | 4 - 5 | 947 | 83 |
AK097728.1 | - | 5 - 6 | 883 | 83 | |
AK301182.1 | - | 2 - 3 | 190 | 83 | |
AY743663.1 | - | 4 - 5 | 1163 | 83 | |
BC038294.2 | - | 4 - 5 | 1163 | 83 |