Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
Chromosome Location
Chr.7: 4644691 - 4771443 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037165.1 NP_001032242.1 3 - 4 915 148
XM_011515191.2 XP_011513493.1 5 - 6 586 148
GenBank mRNA AK122663.1 - 3 - 4 432 148
AK302243.1 - 3 - 4 554 148
BC038434.1 - 6 - 7 659 148

View Full Product Details