The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037165.1 | NP_001032242.1 | 6 - 7 | 1424 | 140 |
XM_011515191.2 | XP_011513493.1 | 8 - 9 | 1095 | 140 | |
GenBank mRNA | AK027883.1 | - | 1 - 2 | 1212 | 140 |
AK122663.1 | - | 7 - 8 | 914 | 140 | |
AK302243.1 | - | 6 - 7 | 1063 | 140 | |
BC038434.1 | - | 9 - 10 | 1168 | 140 |